missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPKAPK3 Recombinant Human Protein
MAPKAPK3 Recombinant Human Protein
Marca: Invitrogen PV3299
Detalles adicionales : Peso : 3.36000kg
Descripción
MAPKAPK3 functions as an integrative element of signaling in both mitogen and stress responses.Especificaciones
NP_004626 | |
Cell Viability, Pharma and Biopharma, Cell Proliferation and Function, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
47.3kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
MAPKAPK3, 3PK, MAPKAP3 | |
MAPKAPK3 was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 2mM DTT, 0.01% NP-40, 500μM ATP, and trace [32P]-γ-ATP for 10 minutes at 30°C | |
MAPKAPK3 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.48mg/mL | |
7867 | |
MAPKAPK3 Recombinant Human Protein | |
–80°C | |
1 Each | |
MAPKAPK3 Recombinant Human Protein | |
MAPKAPK | |
5407nmoles of phosphate transferred to MAPKAP kinase substrate peptide (KKLNRTLSVA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.21μg/mL | |
His Tag | |
Kinase and inhibitors | |
95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.