missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MAPK14 (Human) Recombinant Protein
Human MAPK14 full-length ORF ( AAH00092, 1 a.a. - 360 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001432-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MSQERPTFYRQELNKTIWEVPERYQNLSPVGSGAYGSVCAAFDTKTGLRVAVKKLSRPFQSIIHAKRTYRELRLLKHMKHENVIGLLDVFTPARSLEEFNDVYLVTHLMGADLNNIVKCQKLTDDHVQFLIYQILRGLKYIHSADIIHRDLKPSNLAVNEDCELKILDFGLARHTDDEMTGYVATRWYRAPEIMLNWMHYNQTVDIWSVGCIMAELLTGRTLFPGTDHIDQLKLILRLVGTPGAELLKKISSESARNYIQSLTQMPKMNFANVFIGANPLAVDLLEKMLVLDSDKRITAAQALAHAYFAQYHDPDDEPVADPYDQSFESRDLLIDEWKSLTYDEVISFVPPPLDQEEMES
Especificaciones
AAH00092 | |
mitogen-activated protein kinase 14 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAPK14 | |
GST |
1432 | |
Wheat germ expression system | |
10 μg | |
CSBP1, CSBP2, CSPB1, EXIP, Mxi2, PRKM14, PRKM15, RK, SAPK2A, p38, p38ALPHA | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |