missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP2K2 (MEK2) Recombinant Human Protein
MAP2K2 (MEK2) is a dual specificity serine/threonine kinase.
Marca: Invitrogen™ PR5399A
Detalles adicionales : Peso : 0.01000kg
Descripción
MAP2K2 (MEK2) is known to play a critical role in mitogen growth factor signal transduction.
Especificaciones
NP_109587 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
MAP2K2 (MEK2) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
49.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
STE7 Kinase Family | |
Recombinant | |
Baculovirus |