missing translation for 'onlineSavingsMsg'
Learn More

MAN1C1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-87773

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214437

  • 455.71€ / 100 microlitros
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

MAN1C1 Polyclonal specifically detects MAN1C1 in Mouse samples. It is validated for Western Blot.
Especificaciones

Especificaciones

MAN1C1
Polyclonal
Unconjugated
PBS, 2% Sucrose
Rabbit
Affinity purified
RUO
Primary
Mouse
Purified
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
1,2-alpha-mannosidase IC, Alpha-1,2-mannosidase IC, EC 3.2.1.113, HMICProcessing alpha-1,2-mannosidase IC, MAN1A3, MAN1C, Mannosidase alpha class 1C member 1, mannosidase, alpha, class 1C, member 1, mannosyl-oligosaccharide 1,2-alpha-mannosidase IC, pp6318
The immunogen is a synthetic peptide directed towards the C-terminal region of Mouse MAN1C1
100 μL
Stem Cell Markers
57134
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales