missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluorosilicato de magnesio hidrato, Thermo Scientific Chemicals
Click to view available options
Cantidad:
500 g
2 kg
Descripción
It is employed as an antiplaque agents and oral care agents. Also used for mothproofing textiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Magnesium hexafluorosilicate hydrate |
Punto de fusión | 120°C (decomposition) |
Forma física | Polvo |
Cantidad | 500 g |
Número UN | UN2853 |
Información de solubilidad | Soluble in water. |
Formula Weight (peso de la fórmula) | 166.39 (Anhydrous) |
Densidad | 1.788 g/mL |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'