Learn More
Alfa Aesar™ Magnesium aluminum oxide, 99.9% (metals basis excluding Ca), Ca <25ppm
Marca: Alfa Aesar™ 036388.06
Detalles adicionales : CAS : 12068-51-8 Peso : 0.00500kg
Cantidad. | 5g |
---|
Identificadores de productos químicos
12068-51-8 | |
142.26 | |
UAMZXLIURMNTHD-UHFFFAOYSA-N |
Al2MgO4 | |
MFCD00049475 | |
alumina magnesia, cernel i, dialuminum magnesium tetraoxide, magnesium aluminate mgal2o4, dialuminum;magnesium;oxygen 2-, magnesium aluminate mg alo2 2, aluminum magnesium oxide, aluminum magnesium oxide mgal2o4, magnesium aluminum oxide mgal2o4, aluminate alo21-, magnesium 2:1 |
Especificación
Magnesium aluminum oxide | |
(metals basis excluding Ca), Ca <25ppm | |
MFCD00049475 | |
UAMZXLIURMNTHD-UHFFFAOYSA-N | |
142.26 | |
99.9% | |
3 to 12mm |
12068-51-8 | |
Al2MgO4 | |
alumina magnesia, cernel i, dialuminum magnesium tetraoxide, magnesium aluminate mgal2o4, dialuminum;magnesium;oxygen 2-, magnesium aluminate mg alo2 2, aluminum magnesium oxide, aluminum magnesium oxide mgal2o4, magnesium aluminum oxide mgal2o4, aluminate alo21-, magnesium 2:1 | |
142.26 | |
Fused Pieces | |
Odorless | |
5g |