Learn More
Brady Magicard™ CR80 Adhesive PVC Card
Product Code. 30067365 Shop All Brady Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 500
Product Code. Color Unit Size
30067365 White Pack of 500
1 options
This item is not returnable. View return policy
Product Code. 30067365 Supplier Brady Supplier No. 322004
  • ONLINE EXCLUSIVE PRICE
    221.00€
    200.00€ / paquete de 500
    Ahorro 21.00€
    9% Descuento
Fecha estimada de envoi: 01-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Plain white PVC Card (CR80) with self adhesive backing.
  • Pre-approved for best quality printing on Magicard Printers.
  • Adhesive back.
  • Pack of 500 cards.
Specifications
Quantity 500 Pk.
Product Type Badge
Alternate Part Number M9007-011
Color White
Material PVC
Message Blank
For Use With (Application) Healthcare and Hospital labelling;General identification;Facility identification;Security identification
Height (English) 3.4 in.
Height (Metric) 86 mm
Thickness 0.575 mm
Width (English) 2.1 in.
Width (Metric) 54 mm
Label Type Cards for Printers
For Use With (Equipment) Magicard 300, Magicard 600, Pronto 100, Prima 8