missing translation for 'onlineSavingsMsg'
Learn More

MAGEA8 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 15045-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16814024

  • 399.00€ / 150 microlitros
Fecha estimada de envío: 03-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Especificaciones

Especificaciones

MAGEA8
Polyclonal
Unconjugated
MAGEA8
Cancer/testis antigen 1.8, CT1.8, MAGE 8 antigen, MAGE8, MAGEA8, melanoma antigen family A, 8, Melanoma associated antigen 8
Rabbit
Antigen Affinity Chromatography
RUO
4107
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P43361
MAGEA8
MAGEA8 Fusion Protein Ag6984
150 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales