Learn More
MafA Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca:  Bio-Techne NB400-137FR
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214063
  • 677.09€ / 100 microlitros
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
MafA Polyclonal specifically detects MafA in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones
Especificaciones
MafA
Polyclonal
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
hMafA, Pancreatic beta-cell-specific transcriptional activator, RIPE3B1, transcription factor MafA, Transcription factor RIPE3b1, V-maf musculoaponeurotic fibrosarcoma oncogene homolog A, v-maf musculoaponeurotic fibrosarcoma oncogene homolog A (avian)
Rabbit
Affinity Purified
RUO
Primary
NB400-137 is specific for MafA protein.
Store at 4C in the dark.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
MAFA
The immunogen recognized by this antibody maps to a region between residue 300 and the C-terminus (residue 359) of mouse v-maf musculoaponeurotic fibrosarcoma oncogene homolog A using the numbering given in entry NP_919331.1 (GeneID 378435).
100 μL
Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators
389692
Human, Mouse