missing translation for 'onlineSavingsMsg'
Learn More

MAD2L1 Mouse anti-Human, Clone: 2F5F12, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66014-1-IG-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817515

  • 109.00€ / 20 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Tight junction proteins (TJPs) belong to a family of membrane-associated guanylate kinase (MAGUK) homologs that are involved in the organization of epithelial and endothelial intercellular junctions. TJPs bind to the cytoplasmic C termini of junctional transmembrane proteins and link them to the actin cytoskeleton.
Especificaciones

Especificaciones

MAD2L1
2F5F12
Unconjugated
MAD2L1
Liquid
MAD2L1
MAD2L1 Fusion Protein Ag17512
Protein A
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1.07 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13257
HSMAD2, MAD2, MAD2 like protein 1, MAD2L1
Mouse
IgG2b
20 μL
-20°C
Primary
4085
Videos
SDS
Documentos
Certificados