Learn More
Abcam Lysis Buffer II
Product Code. 32115372 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 x 5 mL
Unit Size:
5mL
Product Code. Quantity Unit Size
32115372 1 x 5 mL 5mL
1 options
This item is not returnable. View return policy
Product Code. 32115372 Supplier Abcam Supplier No. ab3239481x5mL
 Restricted Item
Fecha estimada de envoi: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Lysis Buffer II (ab323948) is part of the reagents, controls & accessories range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.
Lysis Buffer II (ab323948) is part of the reagents, controls & accessories range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1 x 5 mL
Regulatory Status RUO