Description
- Aluminum
- Variable heating
- Connection cable included
Specifications
Specifications
| Type | Product Shelves |
| Capacity (Metric) | 1590 cm2 |
| No. of Shelves | 1 |
| Material | Aluminum |
| Height (English) | 0.6 in. |
| Height (Metric) | 1.6 cm |
| Length (English) | 8.9 in. |
| Length (Metric) | 22.5 cm |