missing translation for 'onlineSavingsMsg'
Learn More

Lymphotoxin beta R/TNFRSF3 Antibody (4H8 WH2), DyLight 650, Novus Biologicals™

Rat Monoclonal Antibody

Marca:  Novus Biologicals NBP2-80054C

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213226

  • 559.38€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Lymphotoxin beta R/TNFRSF3 Monoclonal antibody specifically detects Lymphotoxin beta R/TNFRSF3 in Mouse samples. It is validated for Flow Cytometry
Especificaciones

Especificaciones

Lymphotoxin beta R/TNFRSF3
Monoclonal
DyLight 650
CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III
Recombinant mouse Lymphotoxin beta R/TNFRSF3 (cysteine-rich region/aa 31-221).
0.1 mL
Apoptosis, Cytokine Research
4055
Store at 4°C in the dark.
IgG2a
Flow Cytometry
4H8 WH2
50mM Sodium Borate
Rat
>95%, by SDS-PAGE
RUO
Primary
Mouse
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales