Learn More
Lycopsamine 100 μg/mL in Water, Dr. Ehrenstorfer
Product Code. 30320455
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity Unit Size
30320455 1 mL 1mL
1 options
This item is not returnable. View return policy
Product Code. 30320455 Supplier Dr. Ehrenstorfer Supplier No. DREA14653500WA100
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Lycopsamine
CAS 10285-07-1
Analyte or Component Names Lycopsamine
Shipping Condition +20°C
Molecular Formula C15 H25 N O5
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Butanoic acid, 2,3-dihydroxy-2-(1-methylethyl)-, (2,3,5,7a-tetrahydro-1-hydroxy-1H-pyrrolizin-7-yl)methyl ester, [1R-[1α,7(2S*,3S*),7aβ]]-, Lycopsamine (7CI,8CI), (+)-Lycopsamine, 3'-epi-Intermedine, Retronecine 9-((-)-viridiflorate)
InChI Formula InChI=1S/C15H25NO5/c1-9(2)15(20,10(3)17)14(19)21-8-11-4-6-16-7-5-12(18)13(11)16/h4,9-10,12-13,17-18,20H,5-8H2,1-3H3/t10-,12+,13+,15-/m0/s1
SMILES CC(C)[C@](O)([C@H](C)O)C(=O)OCC1=CCN2CC[C@@H](O)[C@@H]12
IUPAC Name [(7R,8R)-7-hydroxy-5,6,7,8-tetrahydro-3H-pyrrolizin-1-yl]methyl (2S)-2-hydroxy-2-[(1S)-1-hydroxyethyl]-3-methylbutanoate
Solvent or Matrix Water
Molecular Weight (g/mol) 299.363
Quantity 1 mL
Formula Weight 299.173 g/mol