Learn More
Ly-6G Rat, anti-Mouse, Biotin, Clone: 1A8, STEMCELL Technologies
Rat Monoclonal Antibody
Marca:  Stemcell Technologies 60031BT.1
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17198631
  • 126.35€ / 50 microgramos
Fecha estimada de envío: 10-11-2022
para ver el stock
Descripción
Descripción
  • The 1A8 antibody reacts with Ly-6G, a 21 - 25 kDa GPI-anchored protein, which together with the structurally related Ly-6C protein comprises the granulocyte receptor-1 antigen (Gr-1)
  • Gr-1 is expressed on monocytes, neutrophils and subsets of macrophages, plasmacytoid dendritic cells, and T cells
  • Monocytes in the bone marrow transiently express Gr-1 during development and the expression level is strongly correlated with granulocyte differentiation and maturation
  • In the periphery, Gr-1 is found predominantly on neutrophils and is a useful marker for these cells
  • The 1A8 antibody binds specifically to Ly-6G, whereas another commonly used antibody, clone RB6-8C5, binds to both Ly-6G and Ly-6C
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Neutrophil Enrichment Kit (Catalog #19762)
Especificaciones
Especificaciones
Ly-6G
Monoclonal
Biotin
50 μg
Mouse
Granulocytes and Subsets, Myeloid Cells
Functional Assay, Immunocytochemistry, Flow Cytometry, Immunoprecipitation
1A8
Rat
Primary
IgG2a κ