missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Luciferase Rabbit anti-Renilla, Tag, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27986-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody detects the Luciferase protein of Photinus pyralis (Common eastern firefly) (Lampyris pyralis).
Luciferase produces green light with the wavelength of 562 nm. Luciferase catalyzes the reaction: D-firefly luciferin + O2 + ATP = firefly oxyluciferin + CO2 + AMP + diphosphate + light.Especificaciones
Luciferase | |
Polyclonal | |
Unconjugated | |
Luciferase, Luciferin 4-monooxygenase | |
Luciferase Fusion Protein Ag27633 | |
150 μL | |
Primary | |
-20°C | |
Liquid | |
Tag |
Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
Renilla | |
Antibody | |
IgG |