Learn More
Lubricating Oil Standard 5: Ca @ 0.300; P @ 0.060 S @ 0.100; Zn @ 0.130 wt% in Lubricating Oil, VHG™
Product Code. 17951156
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity Unit Size
17951156 100 mL 100mL
1 options
This item is not returnable. View return policy
Product Code. 17951156 Supplier VHG Labs Supplier No. VHGLOIL5100
 Restricted Item
Fecha estimada de envoi: 12-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lubricating Oil Standard 5: Ca @ 0.300; P @ 0.060 S @ 0.100; Zn @ 0.130 wt% in Lubricating Oil
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phosphorus/Sulfur/Zinc/Calcium
CAS 7723-14-0, 7704-34-9, 7440-66-6, 7440-70-2
Methods XRF
Analyte or Component Names Phosphorus/Sulfur/Zinc/Calcium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 20 cSt Mineral Oil
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Phosphorus 0.0604%/Sulfur 0.1004%/Zinc 0.1306%/Calcium 0.3%