Learn More
LST1 Antibody (LST1/02) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NBP1-45072
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18215985
  • 410.36€ / 0.10 miligramo
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
LST1 Monoclonal specifically detects LST1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Especificaciones
Especificaciones
LST1
Monoclonal
1 mg/mL
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:50
LST1
KLH-fused peptide corresponding to amino acids SSEGPDLRGRDKRGT of human LST1
Protein A purified
RUO
Primary
The antibody LST1/02 reacts with an extracellular epitope of LST1, an approximately 6-11 kDa protein expressed as various transmembrane or soluble forms. LST1 is found predominantly on monocytes and dendritic cells. It migrates on SDS PAGE gels as approximately 25-28 kDa molecule.
Store at 4C. Do not freeze.
IgG1
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
LST1/02
Unconjugated
B144D6S49ELST-1, leucocyte specific transcript 1, leukocyte specific transcript 1, leukocyte-specific transcript 1 protein, lymphocyte antigen 117, MGC119006, MGC119007, Protein B144
Mouse
11 kDa
0.1 mg
Cell Cycle and Replication
7940
Human
Purified