missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26119-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
LSM11 is a component of the U7 snRNP complex that is involved in the histone 3′-end pre-mRNA processing. Increases U7 snRNA levels but not histone 3′-end pre-mRNA processing activity, when overexpressed. Required for cell cycle progression from G1 to S phases. Binds specifically to the Sm-binding site of U7 snRNA.Especificaciones
LSM11 | |
Polyclonal | |
Unconjugated | |
LSM11 | |
LSM11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
134353 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P83369 | |
LSM11 | |
LSM11 Fusion Protein Ag23281 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |