missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10874-1-AP-150UL
Additional Details : Peso : 0.00010kg
Description
LSM10 gene ontology annotations related to this gene include positive regulation of G1/S transition of mitotic cell cycle.Specifications
LSM10 | |
Polyclonal | |
Unconjugated | |
LSM10 | |
LSM10, MST074, MSTP074 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116748, 366468, 84967 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8QZX5, Q969L4 | |
Lsm10 | |
LSM10 Fusion Protein Ag1027 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |