missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPHN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20045-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody recognizes all the isoforms of LPHN3 and has no cross reaction with LPHN1 and LPHN2.
Plays a role in cell-cell adhesion and neuron guidance via its interactions with FLRT2 and FLRT3 that are expressed at the surface of adjacent cells. Plays a role in the development of glutamatergic synapses in the cortex. Important in determining the connectivity rates between the principal neurons in the cortex.Especificaciones
LPHN3 | |
Polyclonal | |
Unconjugated | |
ADGRL3 | |
CIRL 3, CIRL3, KIAA0768, latrophilin 3, LEC3, Lectomedin 3, LPHN3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170641, 23284, 319387 | |
-20°C | |
Liquid |
Immunoprecipitation, Flow Cytometry, Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80TS3, Q9HAR2, Q9Z173 | |
Adgrl3 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |