Learn More
Thermo Scientific Alfa Aesar Low Form Inconel Crucible,Cap (ml), 20,Top Dia (mm), 33,Bottom Dia (mm), 25,Depth (mm), 30 1each
Marca: Thermo Scientific Alfa Aesar 035907.KT
Low Form Inconel Crucible,Cap (ml), 20,Top Dia (mm), 33,Bottom Dia (mm), 25,Depth (mm), 30 1each
Detalles adicionales : Peso : 0.01000kg