Learn More
Corning™ Low Base Cyclic Olefin Copolymer Microplate
Low base designed for optimal bottom reader sensitivity. The Low Base Cyclic Olefin Copolymer Microplate has excellent signal dynamic range.
Marca:  Corning™ 4677
Detalles adicionales : Peso : 11.50000kg
 Ver más versiones de este producto
Código de producto. 15883885
  • 4910.65€ / paquete de 100
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • No logo or lettering for improved heat sealing and ease of liquid handling
  • Low base designed for optimal bottom reader sensitivity
  • Optically clear flat well bottom permits direct microscopic viewing
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Superior flatness minimizes variability within a plate and between plates
  • Flat bottom wells are compatible with automated dispensers including the PerkinElmer FlexDrop™
Especificaciones
Especificaciones
Yes
Black
1536
12.5μL
Clear
20
Non-sterile
Flat
Cyclic Olefin Copolymer
100
Up to 8μL
Not Included
Poly-D-Lysine Treated
Videos