Learn More
Lonza™ BioWhittaker™ X-VIVO™ 10 Hematopoietic Serum-Free Culture Media
Product Code. 11675120
Change view
Click to view available options
Quantity:
1L
Unit Size:
1 L
Product Code. Quantity Unit Size
11675120 1L 1 L
1 options
This item is not returnable. View return policy
Product Code. 11675120 Supplier Lonza™ Supplier No. 04380Q
  • ONLINE EXCLUSIVE PRICE
    357.00€
    278.83€ / 1 litros
    Ahorro 78.17€
    21% Descuento
 Restricted Item
Fecha estimada de envoi: 04-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
For adoptive immunotherapy and cellular therapy research
  • Does not contain any lymphokines required for activation of human lymphocytes
  • Intended for in vitro procedures
  • Device Master Records (DMRs) for these products are held in an FDA Master File
  • Chemically defined media optimized for specific cell types
  • For further manufacturing or laboratory use
  • Designed to support the generation of lymphokine activated killer (LAK) cells in a serum-free environment
  • Limited by Federal (USA) law to investigational use
  • These products are for research use only
  • Not approved for human or veterinary use, for application to humans or animals, or for use in clinical or in vitro procedures
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stem Cell Serum Free Media (SFM)
With Additives Phenol Red
Quantity 1L
Content And Storage 2° to 8°C