missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LONP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18035-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
ATP-dependent serine protease that mediates the selective degradation of misfolded and unassembled polypeptides in the peroxisomal matrix. Necessary for type 2 peroxisome targeting signal (PTS2)-containing protein processing and facilitates peroxisome matrix protein import.Especificaciones
LONP2 | |
Polyclonal | |
Unconjugated | |
LONP2 | |
lon peptidase 2, peroxisomal, Lon protease 2, Lon protease like protein 2, LONP, LONP2, LONPL, Peroxisomal Lon protease | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
66887, 83752 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86WA8, Q9DBN5 | |
Lonp2 | |
LONP2 Fusion Protein Ag12650 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |