Learn More
Loganin, TRC
Product Code. 30475931
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475931 10 mg 10mg
30471343 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30475931 Supplier Toronto Research Chemicals Supplier No. TRCL46940510MG
 Restricted Item
Fecha estimada de envoi: 22-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18524-94-2
Molecular Formula C17 H26 O10
Molecular Weight (g/mol) 390.38
Synonym (1S,4aS,6S,7R,7aS)-1-(β-D-Glucopyranosyloxy)-1,4a,5,6,7,7a-hexahydro-6-hydroxy-7-methylcyclopenta[c]pyran-4-carboxylic Acid Methyl Ester, (-)-Loganin, Loganoside, (1S)-1α-(β-D-Glucopyranosyloxy)-1,4aα,5,6,7,7aα-hexahydro-6α-hydroxy-7α-methylcyclopenta[c]pyran-4-carboxylic Acid Methyl Ester, 7-Hydroxy-6-desoxyverbenalin, NSC 606403
IUPAC Name methyl (1S,4aS,6S,7R,7aS)-6-hydroxy-7-methyl-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-4-carboxylate
SMILES COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]3[C@@H](C)[C@@H](O)C[C@H]13
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C17H26O10/c1-6-9(19)3-7-8(15(23)24-2)5-25-16(11(6)7)27-17-14(22)13(21)12(20)10(4-18)26-17/h5-7,9-14,16-22H,3-4H2,1-2H3/t6-,7+,9-,10+,11+,12+,13-,14+,16-,17-/m0/s1
Formula Weight 390.1526
Chemical Name or Material Loganin