Learn More
Thermo Scientific™ Locator™ Cryogenic Rack and Box Systems
Product Code. 10790520 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
Vials (1600, 100/Box)
Vials (1800, 25/Box)
Vials (3600, 100/Box)
Monitoring Options:
Alarm, Recorder
Ultrasonic Level
Voltage:
220 V
Not Applicable
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Monitoring Options Voltage Unit Size
10790520 Vials (1600, 100/Box) Ultrasonic Level 220 V Each
11825392 Vials (3600, 100/Box) Ultrasonic Level 220 V Each
10393513 Vials (1800, 25/Box) Ultrasonic Level 220 V Each
10579712 Vials (1600, 100/Box) Alarm, Recorder Not Applicable Each
10103362 Vials (3600, 100/Box) Alarm, Recorder Not Applicable Each
10432962 Vials (1800, 25/Box) Alarm, Recorder Not Applicable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10790520 Supplier Thermo Scientific™ Supplier No. CY509105
  • ONLINE EXCLUSIVE PRICE
    7769.41€
    6381.00€ / paquete de 1
    Ahorro 1388.41€
    17% Descuento
 Restricted Item
Fecha estimada de envoi: 16-09-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Retrieve cell samples quickly and easily using these cryogenic rack and box systems.
Retrieve cell samples quickly and easily using Thermo Scientific™ Locator™ Cryogenic Rack and Box Systems, which feature a unique hanging rack and gridded box design, advanced vacuum insulation, unmatched temperature uniformity and an innovative liquid nitrogen monitor.
Features:
  • Compact design minimizes space requirements while maximizing storage capacity.
  • System is suited for both manual and computerized inventory record-keeping methods.
  • Super-insulated vessels have special, narrow-mouth design that conserves liquid nitrogen by minimizing evaporation.
  • Samples are stored below -180°C, even when less than 2' (5 cm) of liquid nitrogen remains in the vessel.
  • Ultrasonic Level Monitor safeguards irreplaceable samples with minimal liquid nitrogen evaporation and conduction.
  • Advanced vacuum insulation minimizes liquid nitrogen evaporation and reduces operating costs
  • Optional monitor provides continuous LED readout of liquid nitrogen level; activates visual and audible alarms when level falls below safe range of approximately 1' (2.5 cm).
  • Optional alarm also alerts operator in the event of excessive liquid nitrogen loss.
  • Secure locking hasp prevents unauthorized entry.
Locator Jr.:
  • Static holding time: 70 days
  • 1600 (100/box)
  • Evaporation rate: 0.85 L/day
Locator 4:
  • Static holding time: 112 days
  • 3600 (100/box)
  • Evaporation rate: 0.99 L/day
Locator 8:
  • Static holding time: 185 days
  • 1800 (25/box)
  • Evaporation rate: 0.6 L/day
Includes:
Stainless steel indexed racks that fit into corresponding locations in the vessel. Each rack has individual storage compartments that hold one Thermo Scientific™ Nalgene™ Cryogenic Storage Box.
Requires:
Nalgene Cryogenic Storage Boxes (order separately). Each box has individually numbered storage areas for use with vials sized 2.0 mL and smaller.
Note:
Stated capacities are for Nalgene 1.2/2.0 mL cryogenic vials.
Compatible with:
  • Boxes, Nalgene, 2' for 1.0 and 1.5 mL vials, 100 vials per box, case of 10 (CS509X24)
  • Boxes, Nalgene, 2' for 2 mL vials, 81 vials per box, case of 40 (CS509X4); case of 20 (CS509X5)
  • Boxes, Nalgene, 4' for 5 mL vials, 81 vials per box, each (CS509X10)
  • CryoClaw™ (AY509X18)
  • Cups, Suction, 2 replacements for CryoClaw (AY509X19)
  • Logbook (LT509X5)
  • Level Monitor, customer-installed: CN509X3 (120V, Jr.); CN509X6 (120V, 4); CN509X9 (120V, 8); CN509X1 (220V, Jr.); CN509X4 (220V, 4); CN509X7 (220V, 8)
  • Rack, for 5.0 mL vials (does not include boxes) (HR509X19A)
  • Vials, 1 mL, case of 500 (AY509X32)
  • Vials, Cryogenic, 1.2 mL, case of 500, 1.2 mL (AY509X13)
  • Vials, 1.5 mL, case of 500 (AY509X33)
  • Vials, Cryogenic, 2.0 mL, case of 500, 2.0 mL (AY509X12)
TRUSTED_SUSTAINABILITY
Specifications
Type Locator Jr. with Monitor
Temperature Range Below -180°C
Insulation Vacuum
Certifications/Compliance CE, cCSAus
For Use With (Equipment) Locator™ Cryogenic Rack and Box Systems
Model Locator Jr.
Monitoring Options Ultrasonic Level
Voltage 220 V
Holds Vials (1600, 100/Box)