missing translation for 'onlineSavingsMsg'
Learn More

Perclorato de litio trihidrato, ACS, 63,0-68,0 % LiClO4, Thermo Scientific Chemicals

Catalog Number 15457125
Click to view available options
Cantidad:
10 g
100 g
500 g
Este artículo no se puede devolver. Vea la política de devoluciones
13453-78-6
ClH6LiO7
160.431
MFCD00149765
JAWYRNYHJJDXHX-UHFFFAOYSA-M
lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion
23708903
litio;perclorato;trihidrato
[Li+].O.O.O.[O-]Cl(=O)(=O)=O
Este artículo no se puede devolver. Vea la política de devoluciones
13453-78-6
ClH6LiO7
160.431
MFCD00149765
JAWYRNYHJJDXHX-UHFFFAOYSA-M
lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion
23708903
litio;perclorato;trihidrato
[Li+].O.O.O.[O-]Cl(=O)(=O)=O

Lithium perchlorate trihydrate is used as an oxygen source for chemical oxygen generators as it liberates oxygen at high temperatures. Excellent solubility of lithium perchlorate permits its use in Diels-Alder reactions. It is also employed in lithium-ion batteries as an electrolyte and acts as a chaotropic agent to denature proteins. It is used in the Baylis-Hillman reaction as a catalyst in the coupling of alpha, beta-unsaturated carbonyl compounds with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Lithium perchlorate trihydrate
Color Blanco
Forma física cristalino/a/s
Cantidad 10 g
Fórmula lineal LiClO4·3H2O
Número UN UN1481
Índice Merck 14,5539
Formula Weight (peso de la fórmula) 160.45 (106.40 Anhydrous)
Grado ACS
Concentration or Composition (by Analyte or Components) 63.0 to 68.0% LiClO4
Olor Odorless
Densidad 1.84g/mL
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'