missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Perchlorate, purum p.a, ≥98.0% (calc. based on dry substance, T), Honeywell™
Click to view available options
Cantidad:
100 g
500 g
Envase:
Poly Bottle
Descripción
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Especificaciones
Nombre del producto químico o material | Lithium perchlorate |
Color | White |
pH | 6.0 to 9.5 (at 104g/L 25°C) |
Forma física | Granules |
Cantidad | 100 g |
Fórmula lineal | LiClO4 |
Número UN | UN1481 |
Formula Weight (peso de la fórmula) | 106.39g/mol |
Porcentaje de pureza | ≥98.0% (calc. based on dry substance, T) |
Envase | Poly Bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'