Learn More
Invitrogen™ Human LIS1 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0536
Additional Details : Peso : 0.01000kg
Description
LIS1 Synthetic Peptide, PEP-0536, from Invitrogen.
This peptide corresponds to 14 amino acids near the carboxy terminus of human LIS1.PEP-0536 can be used as a blocking peptide with polyclonal antibody PA5-20419.Specifications
5048 | |
purified | |
Human LIS1 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
P43034 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
PAFAH1B1 | |
0.02% sodium azide | |
50 μg | |
PBS |