missing translation for 'onlineSavingsMsg'
Learn More

Lipocalin-2/NGAL Antibody (17), HRP, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP1-30005H

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214291

  • 788.32€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Porcine samples. It is validated for Western Blot, ELISA.
Especificaciones

Especificaciones

Lipocalin-2/NGAL
Monoclonal
HRP
PBS with No Preservative
LCN2
Recombinant pig NGAL adsorbed onto aluminum hydroxide gel
0.1 mL
Lipid and Metabolism
3934
Store at 4C in the dark.
IgG1 κ
Western Blot, ELISA
17
Western Blot, ELISA
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Mouse
Protein A purified
RUO
Primary
Porcine
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales