missing translation for 'onlineSavingsMsg'
Learn More

Lipocalin-2/NGAL Antibody (17), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP1-30005AF594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213663

  • 668.45€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Porcine samples. It is validated for Western Blot, ELISA.
Especificaciones

Especificaciones

Lipocalin-2/NGAL
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
LCN2
Recombinant pig NGAL adsorbed onto aluminum hydroxide gel
0.1 mL
Primary
Porcine
IgG1 κ
Western Blot, ELISA
17
Western Blot, ELISA
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Mouse
Protein A or G purified
Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience
3934.0
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales