Learn More
Abeba™ Light 31138 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marca: Abeba™ 31138/39
Detalles adicionales : Peso : 0.90000kg
Descripción
- CE EN ISO 20347:2012 O2 FO SRA
- Smooth leather
- Black
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3559)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
Especificaciones
CE EN ISO 20347:2012 O2 FO SRA | |
Leather | |
Slip resistant | |
31138 |
Black | |
39 | |
Polyurethane |