missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP has been found elevated in the serum of patients with cancer and in those infected by the human immunodeficiency virus (HIV). It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Using fluorescence in situ hybridization the full length 90K cDNA has been localized to chromosome 17q25. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1.
Specifications
Specifications
| Antigen | LGALS3BP |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | LGALS3BP |
| Gene Accession No. | Q08380 |
| Gene Alias | 90K, BTBD17B, Galectin 3 binding protein, LGALS3BP, M2BP, Mac 2 binding protein, MAC 2 BP, MAC2BP, Tumor associated antigen 90K |
| Gene Symbols | LGALS3BP |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?