Learn More
Invitrogen™ LentiArray™ Human Cancer Biology CRISPR Library
Product Code. 17285976
Change view
Click to view available options
Quantity:
1 library
Unit Size:
Each
Product Code. Quantity Unit Size
17285976 1 library Each
1 options
This item is not returnable. View return policy
Product Code. 17285976 Supplier Invitrogen™ Supplier No. A42268
 Restricted Item
Fecha estimada de envoi: 23-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
The Invitrogen™ LentiArray™ Human Cancer Biology Library targets 510 genes commonly involved in the development of cancer with up to 4 gRNA per gene target (pooled in a single well) for a total of 2,040 gRNAs. Libraries are delivered as 100 μL of ready to use lentiviral particles per gene target.
The award-winning Invitrogen™ LentiArray™ Human Cancer Biology Library targets 510 genes commonly involved in the development of cancer with up to 4 gRNA per gene target (pooled in a single well) for a total of 2,040 gRNAs. Libraries are delivered as 100 μL of ready to use lentiviral particles per gene target with an average titer of 1x108 TU/mL (functional titer determined by antibiotic resistance).
The gene targets within this library were selected using the most up-to-date genome databases, including the NCBI RefSeq database, and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC), as well as The Cancer Genome Atlas (TCGA).
Built to bring you success
LentiArray CRISPR libraries are constructed using gRNA designs produced using Thermo Fisher Scientific’s proprietary CRISPR gRNA design algorithm. This algorithm incorporates the latest gRNA design research, as well as our extensive in-house experience, to produce the highest quality designs. The gRNA designs included in the LentiArray CRISPR libraries are selected for maximal editing efficiency and specificity and are designed to knock out all known isoforms of the target gene. For each gene target, up to four high-quality gRNAs are included to help ensure highly efficient knockout of the target gene in a wide array of cell types.
Design and execute your experiments without limitations
LentiArray CRISPR libraries are designed and constructed to give you complete control over your experimental design. They utilize a two-vector design, expressing the Cas9 nuclease and the gRNA off of separate lentiviral constructs, enabling you to dictate when and how the genome editing tools are delivered to your cells.
The LentiArray Cas9 lentivirus construct expresses a human, codon-optimized Cas9 nuclease with a blasticidin-resistance gene under the control of the EF-1α (EFS) promoter. You can choose to co-infect your cells with both the LentiArray gRNA lentiviruses and the Cas9 lentivirus or just the LentiArray Cas9 lentivirus to establish a stable Cas9 expressing cell line. With LentiArray CRISPR libraries you have the freedom to design the experimental approach that best suits your model system and your screening goals.
Furthermore, to minimize restrictions to your experimental design, the gRNA lentiviruses that make up the LentiArray CRISPR libraries do not contain fluorescent markers. This allows reporters of any wavelength to be used with the libraries and expands your ability to perform multiplexed experiments. For challenging cell lines, the LentiArray gRNA lentiviruses contain a puromycin resistance gene that allows enrichment of the population of edited cells and drives stronger phenotypes.
Optimize your assay design and have full confidence in your results with LentiArray controls
High quality controls are the cornerstone of a successful screen. The LentiArray CRISPR libraries product line includes a suite of controls to help you quickly develop your assay and give you confidence in defining your hit criteria.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 1 library set, 100 μL/target.
Store at -68 to -85°C.
Delivery Type Lentiviral
Product Line LentiArray
Promoter U6
Quantity 1 library
Shipping Condition Dry Ice