missing translation for 'onlineSavingsMsg'
Learn More

LDLR Antibody (301), Alexa Fluor™ 594, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-89351AF594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215746

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

LDLR Monoclonal specifically detects LDLR in Human samples. It is validated for Flow Cytometry.
Especificaciones

Especificaciones

LDLR
Monoclonal
Alexa Fluor 594
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Flow Cytometry
301
Flow Cytometry
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
This antibody was obtained from a rabbit immunized with purified, recombinant Human LDLR (NP_000518.1; Met1-Arg788).
0.1 mL
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
3949
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales