missing translation for 'onlineSavingsMsg'
Learn More

LDLR Antibody (032), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90523F

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213898

  • 605.82€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

LDLR Monoclonal specifically detects LDLR in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

LDLR
Monoclonal
FITC
PBS
Rabbit
Protein A purified
RUO
Primary
No cross-reactivity in ELISA with: Human LDLR/ LDL Receptor
Store at 4°C in the dark.
IgG
Flow Cytometry, ELISA, Immunofluorescence
32
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LDLR (Met 1-Arg 790; NP_034830.2).
0.1 mL
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
3949
Mouse
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales