missing translation for 'onlineSavingsMsg'
Learn More

LCE1A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 24956-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16813485

  • 319.00€ / 150 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

LCE1A belongs to the late cornified envelope (LCE) gene cluster within the epidermal differentiation complex (EDC) on chromosome 1. The LCE cluster contains multiple conserved genes that encode stratum corneum proteins, and these genes are expressed relatively late during fetal assembly of the skin cornified envelope.
Especificaciones

Especificaciones

LCE1A
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5T7P2
late cornified envelope 1A, Late envelope protein 1, LCE1A, LEP1
Rabbit
IgG
150 μL
-20°C
Primary
353131
Immunohistochemistry (Paraffin)
Unconjugated
LCE1A
Liquid
LCE1A
LCE1A Fusion Protein Ag21047
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documentos
Certificados