missing translation for 'onlineSavingsMsg'
Learn More

LBP Mouse anti-Human, Clone: 3A7E3, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66181-1-IG-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817825

  • 399.00€ / 150 microlitros
Fecha estimada de envío: 17-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The LBP protein is involved in the acute-phase immunologic response to gram-negative bacterial infections. Gram-negative bacteria contain a glycolipid, lipopolysaccharide (LPS), on their outer cell wall. Together with bactericidal permeability-increasing protein (BPI), the encoded protein binds LPS and interacts with the CD14 receptor, probably playing a role in regulating LPS-dependent monocyte responses. This protein is part of a family of structurally and functionally related proteins, including BPI, plasma cholesteryl ester transfer protein (CETP), and phospholipid transfer protein (PLTP). Finally, this gene is found on chromosome 20, immediately downstream of the BPI gene.
Especificaciones

Especificaciones

LBP
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P18428
LBP
LBP Fusion Protein Ag13485
150 μL
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
3A7E3
Unconjugated
LBP
LBP
Mouse
Protein A
RUO
3929
-20°C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales