Learn More
LAP1 Antibody (RL13), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NB120-2737AF647
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18216220
  • 551.82€ / 0.20 ml
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
LAP1 Monoclonal specifically detects LAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
LAP1
Monoclonal
Alexa Fluor 647
lamina-associated polypeptide 1B, torsin A interacting protein 1
Mouse
Protein A or G purified
RUO
Primary
Detects lamin associated polypeptide (LAP) 1A, 1B & 1C from rat tissues. This does not cross-react with LAP 2.This detects 3 distinct proteins of molecular weights 75, 68 and 55 kDa representing LAP 1A, 1B and 1C, respectively in rat liver nuclear envelope fractions. Immunofluorescence staining of LAP 1 in rat liver with NB120-2737 results in exclusive labeling of the nuclear periphery and exhibits co-localization with lamin staining.
Store at 4C in the dark.
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
RL13
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
TOR1AIP1
Pore complex-lamina fraction isolated from rat liver nuclear envelopes.
0.2 mL
Apoptosis, Immunology, Innate Immunity, Zinc Finger
208263
Mouse, Rat
Purified