Learn More
Brady™ Laminated Polyester Not Drinking Water Safety Sign
Product Code. 13274359 Shop All Brady Products
Change view
Click to view available options
Material:
Laminated Polyester
Unit Size:
Pack of 2
Product Code. Material Unit Size
13274359 Laminated Polyester Pack of 2
1 options
This item is not returnable. View return policy
Product Code. 13274359 Supplier Brady™ Supplier No. 822441
  • ONLINE EXCLUSIVE PRICE
    5.05€
    4.89€ / paquete de 2
    Ahorro 0.16€
    3% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC P005-Dia 050-PE-SHEET/2
Finish Gloss
Material Laminated Polyester
Quantity 2/Pk.
Adhesive Type Permanent Acrylic
For Use With (Application) Indoor & Outdoor
Thickness 0.099 mm
Temperature Range (Metric) -40 °C-110 °C
Shape Round
Languages Global
Color Red