missing translation for 'onlineSavingsMsg'
Learn More

Lambda Light Chain Antibody (4C2), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP1-45071APCCY7

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215745

  • 521.58€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry.
Especificaciones

Especificaciones

Lambda Light Chain
Monoclonal
APC-Cyanine7
PBS with 0.05% Sodium Azide
IGLC1
The antibody 4C2 reacts with lambda light chains (22.5 kDa) of human immunoglobulin.
0.1 mL
Primary
The mouse monoclonal antibody H4B4 recognizes CD107b/ LAMP-2, an extensively glycosylated 100-120 kDa widely expressed lysosome-associated protein.
Store at 4°C in the dark. Do not freeze.
Flow Cytometry
4C2
Flow Cytometry
IGLC, immunoglobulin lambda constant group
Mouse
Protein A purified
Adaptive Immunity, Immunology
3537.0
Human
IgG1
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales