missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ LAMA4 (Human) Recombinant Protein
Human LAMA4 full-length ORF ( AAH04241.1, 1 a.a. - 120 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00003910-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MALSSAWRSVLPLWLLWSAACSRAASGDDNAFPFDIEGSSAVGRQDPPETSEPRVALGRLPPAAEVQCPCHCHPAGAPAPPRAVPHSSFSLSPPLSSPQCLESFTWARSVRKLEIKSFPL
Especificaciones
AAH04241.1 | |
laminin, alpha 4 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LAMA4 | |
GST |
3910 | |
Wheat germ expression system | |
25 μg | |
DKFZp686D23145, LAMA3, LAMA4*-1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |