Learn More
Melford Lactic Acid 88% Solution
Product Code. 30018371
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Unit Size:
1 L
4 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018371 500 mL 500mL
30018370 4 L 4 L
30018369 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018371 Supplier Melford Supplier No. L17500500.0
  • ONLINE EXCLUSIVE PRICE
    56.30€
    37.73€ / 500 ml
    Ahorro 18.57€
    32% Descuento
 Restricted Item
Fecha estimada de envoi: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
ACS Reagent Grade
ACS Reagent Grade
Specifications
Chemical Name or Material Lactic Acid
CAS 50-21-5
Purity Grade Notes ACS Reagent Grade
Molecular Formula C3H6O3
Quantity 500 mL