missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D System, ASI 26/77 film, 2 Panel Ports

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

Marca:  Thermo Scientific™ SS00105-I

Detalles adicionales : Peso : 4.90000kg

 Ver más versiones de este producto

Código de producto. 15929926

  • 335.67€ / 5 unidades


Fecha de envío estimada
del proveedor 19-12-2019
Añadir a la cesta

Descripción

Descripción

This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 2 ports (1 MPC insert + cap, 1 MPC body + cap). It is available in 50, 100 and 200L capacities.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors

BPC port specifications:

  • Line 1: MPC insert with 9.5mm (3/8 in.) barb, cap, and pinch clamp. Tubing: Silicone, length: 51cm (20 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
  • Line 2: MPC body with 9.5mm (3/8 in.) barb, plug, and pinch clamp.

Especificaciones

Especificaciones

Polyethylene
 ASI 26/77
This product is packaged and sealed into plastic dust cover packaging which is then placed into carboard cartons labeled with product and lot identification.
Labtainer 2D BioProcess Containter System
Irradiated
C-Flex, 51 cm (20 in.)
142.2 x 95.3 cm (56.0 x 37.5 in.)
TK-0200-05
MPC connections + caps, pinch clamps
5 per case
Store at room temperature.
200 L
Videos
Documentos