missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ BioProcess Container 2D System, ASI 28 Film, 3 Edge Ports, Luer Insert/Luer Body

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

Marca:  Thermo Scientific™ SS00003-I

Detalles adicionales : Peso : 4.20000kg

 Ver más versiones de este producto

Código de producto. 15929896

  • 646.00€ / 50 unidades


Fecha de envío estimada
del proveedor 30-03-2020
Añadir a la cesta

Descripción

Descripción

This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors
Especificaciones

Especificaciones

20.3 x 16.5 cm (8.0 x 6.5 in.)
This product is packaged and sealed into plastic dust cover packaging which is then placed into carboard cartons labeled with product and lot identification.
Labtainer 2D BioProcess Containter System
Irradiated
EVA
Ethylene Vinyl Acetate (EVA)
 ASI 28
Luer lock connectors and injection site
50 per case
Store at room temperature.
500 mL
Videos
Documentos