Learn More
SHP Steriltechnik AG Laboklav 195 Trolley
Product Code. 31908773
Change view
Click to view available options
Capacity (Metric):
330 kg
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
31908773 330 kg Each
1 options
This item is not returnable. View return policy
Product Code. 31908773 Supplier SHP Steriltechnik AG Supplier No. 20.0195.000.301
Fecha estimada de envoi: 26-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Trolley for Laboklav 195, horizontal unit.
  • Stainless Steel
  • With 4 castors for easy transport
Specifications
Product Type Trolley for Steam Sterilizer
Capacity (Metric) 330 kg
For Use With (Equipment) Laboklav 195