Learn More
Labconco™ Vacuum Tubing Kit for CentriVap™ System and RapidVap™
Product Code. 10409543 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Polyurethane
Unit Size:
Each
Product Code. Material Unit Size
10409543 Polyurethane Each
1 options
This item is not returnable. View return policy
Product Code. 10409543 Supplier Labconco™ Supplier No. 7814500
  • ONLINE EXCLUSIVE PRICE
    228.00€
    225.05€ / paquete de 1
    Ahorro 2.95€
    1% Descuento
 Restricted Item
Fecha estimada de envoi: 30-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Vacuum Tubing Kit for use with Labconco CentriVap Vacuum Concentrators and RapidVap Evaporators.
Vacuum Tubing Kit for CentriVap Concentrators and RapidVap Evaporators includes two each 1/2 in. ID vacuum tubing, 48 in. length, and 4 tubing clamps.
Specifications
Material Polyurethane
For Use With (Equipment) CentriVap™ system
Type Vacuum Tubing Kit
Weight (English) 6 lb.
Weight (Metric) 2.7 kg