missing translation for 'onlineSavingsMsg'
Learn More

Vascular Endothelial Growth Factor (VEGF) Rabbit Polyclonal Antibody, Epredia™

Take advantage of one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ Vascular Endothelial Growth Factor (VEGF), Rabbit Polyclonal Antibody.

Marca:  Epredia RB-9031-R7

Detalles adicionales : Peso : 0.01200kg

 Ver más versiones de este producto

Código de producto. 12683367

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

VEGF (vascular endothelial growth factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. This antibody reacts with the 165, 189 and 121 amino acid splice variants of VEGF of human, to a lesser extent, mouse and rat.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: N-terminus

Immunogen: A synthetic peptide derived from N-terminal of human VEGF

Molecular Weight: 19-22kDa (reduced)

Positive Control: Angiosarcoma

Cellular Localization: Cytoplasmic, cell surface and extracellular matrix This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation
Especificaciones

Especificaciones

Vascular Endothelial Growth Factor (VEGF)
Polyclonal
Rabbit
7 mL
Angiogenesis
Human
IgG
Immunoprecipitation, Western Blot
Unconjugated
A synthetic peptide derived from N-terminal of human VEGF
IVD
Primary
Prediluted
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales