missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Tubulin-α, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Epredia™ Tubulin-α, Rabbit Polyclonal Antibody.

Marca:  Epredia™ RB-9281-P0

Detalles adicionales : Peso : 0.00170kg

 Ver más versiones de este producto

Código de producto. 12683547

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »


Descripción

Descripción

Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55kDa subunits: α and β. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Cow, Pig, Rat, Mouse, Guinea Pig, Gerbil, Frog and Chicken.

Epitope: aa 426-450

Immunogen: A synthetic peptide derived from aa 426-450 of human tubulin-α

Molecular Weight: 57kDa

Positive Control: MAD109 cells, MCF-7 cells, skin or lung

Cellular Localization: Cytoplasmic This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Especificaciones

Especificaciones

Tubulin-alpha
Polyclonal
Unconjugated
A synthetic peptide derived from aa 426-450 of human tubulin alpha
IVD
Primary
Concentrated
Western Blot
200µg/mL
Rabbit
100 μL
Cytoskeletal Protein
Bovine, Chicken, Frog, Gerbil, Guinea Pig, Human, Mouse, Porcine
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales